3-Iodo-4-nitroanisole, 97%
3-Iodo-4-nitroanisole, 97%
3-Iodo-4-nitroanisole, 97%
3-Iodo-4-nitroanisole, 97%
Thermo Scientific Chemicals

3-Iodo-4-nitroanisole, 97%

CAS: 214279-40-0 | C7H6INO3 | 279.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 326320250
Price (EUR)
274,00
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
274,00
Each
Add to cart
Chemical Identifiers
CAS214279-40-0
IUPAC Name2-iodo-4-methoxy-1-nitrobenzene
Molecular FormulaC7H6INO3
InChI KeyNLTAHTYBZZSCIT-UHFFFAOYSA-N
SMILESCOC1=CC=C(C(I)=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)Yellow to orange-brown
Infrared spectrumConforms
Melting point68°C to 73°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only