Diethyl (chloromethyl)phosphonate, 98%, Thermo Scientific Chemicals
Diethyl (chloromethyl)phosphonate, 98%, Thermo Scientific Chemicals
Diethyl (chloromethyl)phosphonate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl (chloromethyl)phosphonate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 330650010
Price (EUR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3167-63-3
IUPAC Namediethyl (chloromethyl)phosphonate
Molecular FormulaC5H12ClO3P
InChI KeyMZBIWKMCTWJLPT-UHFFFAOYSA-N
SMILESCCOP(=O)(CCl)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.4380 to 1.4400 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only