Methyl-α-D-galactopyranoside, 98+%
Methyl-α-D-galactopyranoside, 98+%
Methyl-α-D-galactopyranoside, 98+%
Thermo Scientific™

Methyl-α-D-galactopyranoside, 98+%

CAS: 3396-99-4 | C7H14O6 | 194.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 337090025
Price (EUR)
62,80
Each
Quantity:
2.5 g
Request bulk or custom format
Price (EUR)
62,80
Each
Chemical Identifiers
CAS3396-99-4
IUPAC Name2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Molecular FormulaC7H14O6
InChI KeyHOVAGTYPODGVJG-UHFFFAOYNA-N
SMILESCOC1OC(CO)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
HPLC>=98.0 %
Appearance of solution(5% in water) Clear colorless
Water7.5 to 9.5 % (K.F.)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only