Methyl Red hydrochloride, ACS reagent
Methyl Red hydrochloride, ACS reagent
Methyl Red hydrochloride, ACS reagent
Thermo Scientific Chemicals

Methyl Red hydrochloride, ACS reagent

CAS: 63451-28-5 | C15H16ClN3O2 | 305.762 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 339320250
Price (EUR)
32,40
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
32,40
Each
Chemical Identifiers
CAS63451-28-5
IUPAC Namehydrogen 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid chloride
Molecular FormulaC15H16ClN3O2
InChI KeyICTWKXAVJAHRMK-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue to dark purple
Visual transition intervalFrom pH 4.2 (pink) to pH 6.2 (yellow)
Appearance (Form)Powder
Infrared spectrumConforms
Clarity of alcohol solutionPasses test
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only