1,3-Dimethylnaphthalene, 96%
1,3-Dimethylnaphthalene, 96%
1,3-Dimethylnaphthalene, 96%
Thermo Scientific Chemicals

1,3-Dimethylnaphthalene, 96%

CAS: 575-41-7 | C12H12 | 156.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3469000101 gGlass bottle
Catalog number 346900010
Price (EUR)
41,10
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
41,10
Each
Chemical Identifiers
CAS575-41-7
SpecificationsSpecification SheetSpecification Sheet
GC>=95 %
Appearance (Color)Clear yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.6080 to 1.6100 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only