Barium peroxide, 95%
Barium peroxide, 95%
Barium peroxide, 95%
Thermo Scientific Chemicals

Barium peroxide, 95%

CAS: 1304-29-6 | BaO2 | 169.33 g/mol
Have Questions?
Catalog NumberQuantityPackaging
350415000500 gPlastic bottle
Catalog number 350415000
Price (EUR)
95,65
線上優惠
106,00
Save 10,35 (10%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
95,65
線上優惠
106,00
Save 10,35 (10%)
Each
Chemical Identifiers
CAS1304-29-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Iodometric>=94 %
Appearance (Color)Beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only