4-Borono-L-phenylalanine, 97%
4-Borono-L-phenylalanine, 97%
4-Borono-L-phenylalanine, 97%
Thermo Scientific Chemicals

4-Borono-L-phenylalanine, 97%

CAS: 76410-58-7 | C9H12BNO4 | 209.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
250 mg
Packaging:
Glass bottle
Catalog number 358932500
Price (EUR)
158,00
Each
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
158,00
Each
Chemical Identifiers
CAS76410-58-7
IUPAC Name(2S)-2-amino-3-[4-(dihydroxyboranyl)phenyl]propanoic acid
Molecular FormulaC9H12BNO4
InChI KeyNFIVJOSXJDORSP-QMMMGPOBSA-N
SMILESN[C@@H](CC1=CC=C(C=C1)B(O)O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only