Silica gel, C18-RP, 17%C, approx. 0.7 mmol/g, part. size: 48-65 micron
Silica gel, C18-RP, 17%C, approx. 0.7 mmol/g, part. size: 48-65 micron
Silica gel, C18-RP, 17%C, approx. 0.7 mmol/g, part. size: 48-65 micron
Thermo Scientific Chemicals

Silica gel, C18-RP, 17%C, approx. 0.7 mmol/g, part. size: 48-65 micron

CAS: 71889-02-6 | C18H37Cl3O2Si2 | 448.012 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass Bottle
Catalog number 360120500
Price (EUR)
233,00
Each
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
233,00
Each
Chemical Identifiers
CAS71889-02-6
IUPAC Namesilanedione; trichloro(octadecyl)silane
Molecular FormulaC18H37Cl3O2Si2
InChI KeyVYTRPUAMVCWPLO-UHFFFAOYSA-N
SMILESO=[Si]=O.CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Carbon (C)>=16 %
Volatile matter=<4 % (110°C)
Specific surface area500 to 600 m²/g
Pore volume0.80 to 1.00 ml/g
Particle size48.2 to 65.3 µm (D50)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
RUO – Research Use Only