Palladium(II) chloride, 99.999%, (trace metal basis)
Palladium(II) chloride, 99.999%, (trace metal basis)
Palladium(II) chloride, 99.999%, (trace metal basis)
Thermo Scientific Chemicals

Palladium(II) chloride, 99.999%, (trace metal basis)

CAS: 7647-10-1 | Cl2Pd | 177.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 369670010
Price (EUR)
208,00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
208,00
Each
Chemical Identifiers
CAS7647-10-1
IUPAC Namepalladium(2+) dichloride
Molecular FormulaCl2Pd
InChI KeyPIBWKRNGBLPSSY-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Total trace metal impurities=<0.001 %
Appearance (Form)Powder
Assay59 to 61 % (Pd)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only