tert-Butyl 4-amino-1-piperidinecarboxylate, 98%
tert-Butyl 4-amino-1-piperidinecarboxylate, 98%
tert-Butyl 4-amino-1-piperidinecarboxylate, 98%
Thermo Scientific Chemicals

tert-Butyl 4-amino-1-piperidinecarboxylate, 98%

CAS: 87120-72-7 | C10H21N2O2 | 201.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 384860050
Price (EUR)
76,40
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
76,40
Each
Chemical Identifiers
CAS87120-72-7
IUPAC Name1-[(tert-butoxy)carbonyl]piperidin-4-aminium
Molecular FormulaC10H21N2O2
InChI KeyLZRDHSFPLUWYAX-UHFFFAOYSA-O
SMILESCC(C)(C)OC(=O)N1CCC([NH3+])CC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks
Appearance (Color)White to light yellow to beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only