Rhodium(II) trifluoroacetate dimer
Rhodium(II) trifluoroacetate dimer
Rhodium(II) trifluoroacetate dimer
Thermo Scientific Chemicals

Rhodium(II) trifluoroacetate dimer

CAS: 31126-95-1 | C8F12O8Rh2 | 657.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 389700010
Price (EUR)
544,00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
544,00
Each
Chemical Identifiers
CAS31126-95-1
IUPAC Nametetrakis(trifluoroacetic acid) dirhodium
Molecular FormulaC8H4F12O8Rh2
InChI KeySZQVMUPTZFMHQT-UHFFFAOYSA-N
SMILES[Rh].[Rh].OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.OC(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Rhodium (Rh)27.5 to 35.0 %
Appearance (Color)Green
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only