2,5-Dibromothiazole, 97%
2,5-Dibromothiazole, 97%
2,5-Dibromothiazole, 97%
Thermo Scientific Chemicals

2,5-Dibromothiazole, 97%

CAS: 4175-78-4 | C3HBr2NS | 242.916 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 397100010
Price (EUR)
37,10
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
37,10
Each
Chemical Identifiers
CAS4175-78-4
IUPAC Name2,5-dibromo-1,3-thiazole
Molecular FormulaC3HBr2NS
InChI KeyXIBIQFJKUZZLLX-UHFFFAOYSA-N
SMILESBrC1=CN=C(Br)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point45°C to 50°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only