D-Valine methyl ester hydrochloride, 99%
D-Valine methyl ester hydrochloride, 99%
D-Valine methyl ester hydrochloride, 99%
Thermo Scientific Chemicals

D-Valine methyl ester hydrochloride, 99%

CAS: 7146-15-8 | C6H14ClNO2 | 167.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 398720010
Price (EUR)
26,60
Each
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
26,60
Each
Chemical Identifiers
CAS7146-15-8
IUPAC Namemethyl (2R)-2-amino-3-methylbutanoate hydrochloride
Molecular FormulaC6H14ClNO2
InChI KeyKUGLDBMQKZTXPW-NUBCRITNSA-N
SMILESCl.COC(=O)[C@H](N)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric98.5 to 101.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only