Azetidine hydrochloride, 97%
Azetidine hydrochloride, 97%
Azetidine hydrochloride, 97%
Thermo Scientific Chemicals

Azetidine hydrochloride, 97%

CAS: 36520-39-5 | C3H7N·HCl | 93.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 398730010
Price (EUR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS36520-39-5
IUPAC Nameazetidine hydrochloride
Molecular FormulaC3H8ClN
InChI KeyHGQULGDOROIPJN-UHFFFAOYSA-N
SMILESCl.C1CNC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only