4-Benzyloxy-1-butanol, 97%
4-Benzyloxy-1-butanol, 97%
4-Benzyloxy-1-butanol, 97%
Thermo Scientific Chemicals

4-Benzyloxy-1-butanol, 97%

CAS: 4541-14-4 | C11H16O2 | 180.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 399120250
Price (EUR)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4541-14-4
IUPAC Name4-(benzyloxy)butan-1-ol
Molecular FormulaC11H16O2
InChI KeyTYROJDFHUXSBHC-UHFFFAOYSA-N
SMILESOCCCCOCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5120 to 1.5140 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only