Cresyl Violet perchlorate, 98%, pure
Cresyl Violet perchlorate, 98%, pure
Cresyl Violet perchlorate, 98%, pure
Thermo Scientific Chemicals

Cresyl Violet perchlorate, 98%, pure

CAS: 41830-80-2 | C16H12ClN3O5 | 361.738 g/mol
Have Questions?
Catalog NumberQuantity
405775000500 mg
Catalog number 405775000
Price (EUR)
216,00
Each
Quantity:
500 mg
Request bulk or custom format
Price (EUR)
216,00
Each
Chemical Identifiers
CAS41830-80-2
SpecificationsSpecification SheetSpecification Sheet
UV Lambda max590 to 596 nm (in Methanol)
Appearance (Color)Green
Infrared spectrumConforms
Molar absorption coefficient67400 l/(mol.cm) (at Lambda max)
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only