Quinine sulfate dihydrate, specified according to the requirements of USP
Quinine sulfate dihydrate, specified according to the requirements of USP
Quinine sulfate dihydrate, specified according to the requirements of USP
Quinine sulfate dihydrate, specified according to the requirements of USP
Thermo Scientific Chemicals

Quinine sulfate dihydrate, specified according to the requirements of USP

CAS: 6119-70-6 | C40H54N4O10S | 782.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 418780250
Price (EUR)
279,00
Each
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
279,00
Each
Add to cart
Chemical Identifiers
CAS6119-70-6
IUPAC Namebis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
Molecular FormulaC40H54N4O10S
InChI KeyZHNFLHYOFXQIOW-OIGVVMIYNA-N
SMILESO.O.OS(O)(=O)=O.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.1 % (in mixture of chloroform-alcohol (2:1))
Dihydroquinine sulfate=<10.0 %
Appearance (Color)White
Identification BPasses test
Titration with HClO499.0 to 101.0 % (on anhydrous basis)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only