4-Vinylbenzyl chloride, 90%, technical
4-Vinylbenzyl chloride, 90%, technical
4-Vinylbenzyl chloride, 90%, technical
Thermo Scientific Chemicals

4-Vinylbenzyl chloride, 90%, technical

CAS: 1592-20-7 | C9H9Cl | 152.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 422525000
Price (EUR)
460,65
Online Exclusive
512,00
Save 51,35 (10%)
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
460,65
Online Exclusive
512,00
Save 51,35 (10%)
Each
Chemical Identifiers
CAS1592-20-7
IUPAC Name1-(chloromethyl)-4-ethenylbenzene
Molecular FormulaC9H9Cl
InChI KeyZRZHXNCATOYMJH-UHFFFAOYSA-N
SMILESClCC1=CC=C(C=C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to dark yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=88 %
Refractive index1.5730 to 1.5770 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only