Sodium nitrite, 97+%, ACS reagent
Sodium nitrite, 97+%, ACS reagent
Sodium nitrite, 97+%, ACS reagent
Thermo Scientific Chemicals

Sodium nitrite, 97+%, ACS reagent

CAS: 7632-00-0 | NNaO2 | 69.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2 kg
Catalog number 424350020
Price (EUR)
182,00
Each
Quantity:
2 kg
Request bulk or custom format
Price (EUR)
182,00
Each
Specifications
Assay Percent Range97+%
CAS7632-00-0
ChEBICHEBI:78870
Chemical Name or MaterialSodium nitrite
ColorYellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only