4-[(N-BOC)aminomethyl]aniline, 97%
4-[(N-BOC)aminomethyl]aniline, 97%
4-[(N-BOC)aminomethyl]aniline, 97%
4-[(N-BOC)aminomethyl]aniline, 97%
Thermo Scientific Chemicals

4-[(N-BOC)aminomethyl]aniline, 97%

CAS: 94838-55-8 | C12H18N2O2 | 222.29 g/mol
Have Questions?
Catalog NumberQuantity
4262000101 g
Catalog number 426200010
Price (EUR)
68,30
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
68,30
Each
Add to cart
Chemical Identifiers
CAS94838-55-8
IUPAC Nametert-butyl N-[(4-aminophenyl)methyl]carbamate
Molecular FormulaC12H18N2O2
InChI KeyUXWQXBSQQHAGMG-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NCC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan
Infrared spectrumConforms
Appearance (Form)Crystalline powder
NMR (1H-NMR)Consistent with structure
Melting point73°C to 82°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only