3-Amino-1-adamantanol, 96%
3-Amino-1-adamantanol, 96%
3-Amino-1-adamantanol, 96%
3-Amino-1-adamantanol, 96%
Thermo Scientific Chemicals

3-Amino-1-adamantanol, 96%

CAS: 702-82-9 | C10H17NO | 167.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 435380050
Price (EUR)
173,00
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
173,00
Each
Add to cart
Chemical Identifiers
CAS702-82-9
IUPAC Name3-aminoadamantan-1-ol
Molecular FormulaC10H17NO
InChI KeyDWPIPTNBOVJYAD-UHFFFAOYNA-N
SMILESNC12CC3CC(C1)CC(O)(C3)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental Analysis7.55 to 8.35 % N
Appearance (Form)Crystalline powder
Water=<10 % (K.F.)
Elemental Analysis64.77 to 71.55 % C
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only