4-(Carboxymethyl)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
4-(Carboxymethyl)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
4-(Carboxymethyl)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Carboxymethyl)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
4446700101 g
Catalog number 444670010
Price (EUR)
149,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
149,00
Each
Chemical Identifiers
CAS797755-07-8
IUPAC Name2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid
Molecular FormulaC14H19BO4
InChI KeyFNLWHBHWDXCWHV-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=C(CC(O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH96.0 to 104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only