2-Fluoropyridine-4-boronic acid, 98%
2-Fluoropyridine-4-boronic acid, 98%
2-Fluoropyridine-4-boronic acid, 98%
Thermo Scientific Chemicals

2-Fluoropyridine-4-boronic acid, 98%

CAS: 401815-98-3 | C5H5BFNO2 | 140.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 448730050
Price (EUR)
455,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
455,00
Each
Chemical Identifiers
CAS401815-98-3
IUPAC Name(2-fluoropyridin-4-yl)boronic acid
Molecular FormulaC5H5BFNO2
InChI KeyWXGBZJJAGLSBPR-UHFFFAOYSA-N
SMILESOB(O)C1=CC=NC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=97.5 %
Appearance (Color)Light yellow to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only