Di-tert-butyl phosphite, 96%
Di-tert-butyl phosphite, 96%
Di-tert-butyl phosphite, 96%
Thermo Scientific Chemicals

Di-tert-butyl phosphite, 96%

CAS: 13086-84-5 | C8H18O3P | 193.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 449390250
Price (EUR)
211,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
211,00
Each
Chemical Identifiers
CAS13086-84-5
IUPAC Namebis(tert-butoxy)(oxo)-λ⁵-phosphanylium
Molecular FormulaC8H18O3P
InChI KeyGEBLOQXLELCEEO-UHFFFAOYSA-N
SMILESCC(C)(C)O[P+](=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=95.0 %
Refractive index1.4190 to 1.4210 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only