Sodium phosphate, tribasic dodecahydrate, extra pure
Sodium phosphate, tribasic dodecahydrate, extra pure
Sodium phosphate, tribasic dodecahydrate, extra pure
Thermo Scientific Chemicals

Sodium phosphate, tribasic dodecahydrate, extra pure

CAS: 10101-89-0 | Na3O412H2O | 380.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 450660010
Price (EUR)
34,40
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
34,40
Each
Chemical Identifiers
CAS10101-89-0
IUPAC Nametrisodium dodecahydrate phosphate
Molecular FormulaH24Na3O16P
InChI KeyASTWEMOBIXQPPV-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Free alkali (NaOH)=<3.5 %
Titration excess HCl>=97.5 %
Appearance (Form)Powder or crystals or needles
Insoluble matter=<0.05 % (in water)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only