5-Norbornene-2-carboxylic acid, 98%, mixture of isomers, Thermo Scientific Chemicals
5-Norbornene-2-carboxylic acid, 98%, mixture of isomers, Thermo Scientific Chemicals
5-Norbornene-2-carboxylic acid, 98%, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Norbornene-2-carboxylic acid, 98%, mixture of isomers, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
453301000100 g
Catalog number 453301000
Price (EUR)
249,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
249,00
Each
Chemical Identifiers
CAS120-74-1
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 % (total of isomers)
Titration with NaOH97.5 to 102.5 %
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid or solid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only