Product Image
Thermo Scientific Chemicals

Gemcitabine hydrochloride, 98%

CAS: 122111-03-9 | C9H12ClF2N3O4 | 299.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461060010
Price (EUR)
98,65
Online Exclusive
109,00
Save 10,35 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
98,65
Online Exclusive
109,00
Save 10,35 (9%)
Each
Chemical Identifiers
CAS122111-03-9
IUPAC Namehydrogen 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one chloride
Molecular FormulaC9H12ClF2N3O4
InChI KeyOKKDEIYWILRZIA-OSZBKLCCSA-N
SMILES[H+].[Cl-].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)C1(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
Titration Argentometric97.5 to 102.5 %
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only