Valnemulin hydrochloride
Product Image
Thermo Scientific Chemicals

Valnemulin hydrochloride

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
462832500250 mg
4628300101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 462832500
Price (EUR)
101,00
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
101,00
Each
Add to cart
Chemical Identifiers
CAS133868-46-9
IUPAC Name4-ethenyl-3-hydroxy-2,4,7,14-tetramethyl-9-oxotricyclo[5.4.3.0¹,⁸]tetradecan-6-yl 2-{[1-(2-amino-3-methylbutanamido)-2-methylpropan-2-yl]sulfanyl}acetate hydrochloride
Molecular FormulaC31H53ClN2O5S
InChI KeyMFBPRQKHDIVLOJ-UHFFFAOYNA-N
SMILESCl.CC(C)C(N)C(=O)NCC(C)(C)SCC(=O)OC1CC(C)(C=C)C(O)C(C)C23CCC(=O)C2C1(C)C(C)CC3
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotationon dry substance
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only