Chemical Identifiers
CAS125-46-2
IUPAC Name4,10-diacetyl-11,13-dihydroxy-2,12-dimethyl-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(13),6,9,11-tetraene-3,5-dione
Molecular FormulaC18H16O7
InChI KeyCUCUKLJLRRAKFN-UHFFFAOYNA-N
SMILESCC(=O)C1C(=O)C=C2OC3=C(C(C)=O)C(O)=C(C)C(O)=C3C2(C)C1=O
View more