Product Image
Thermo Scientific Chemicals

Glycitein

CAS: 40957-83-3 | C16H12O5 | 284.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
Catalog number 464640500
Price (EUR)
185,00
Each
Quantity:
50 mg
Request bulk or custom format
Price (EUR)
185,00
Each
Chemical Identifiers
CAS40957-83-3
IUPAC Name7-hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-chromen-4-one
Molecular FormulaC16H12O5
InChI KeyDXYUAIFZCFRPTH-UHFFFAOYSA-N
SMILESCOC1=C(O)C=C2OC=C(C(=O)C2=C1)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
HPLC>=97.5 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only