Product Image
Thermo Scientific Chemicals

N,N-Dimethylsulfamide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 464680010
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS3984-14-3
IUPAC Name(dimethylsulfamoyl)amine
Molecular FormulaC2H8N2O2S
InChI KeyQMHAHUAQAJVBIW-UHFFFAOYSA-N
SMILESCN(C)S(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Melting point95°C to 99°C
GC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only