Product Image
Thermo Scientific Chemicals

Sesamin, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 464892500
Price (EUR)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS607-80-7
IUPAC Name5-[(1S,3aR,4S,6aR)-4-(2H-1,3-benzodioxol-5-yl)-hexahydrofuro[3,4-c]furan-1-yl]-2H-1,3-benzodioxole
Molecular FormulaC20H18O6
InChI KeyPEYUIKBAABKQKQ-AFHBHXEDSA-N
SMILESC1OC2=CC=C(C=C2O1)[C@H]1OC[C@H]2[C@@H]1CO[C@@H]2C1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Color)White to light yellow
Melting point121°C to 125°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only