Azetidine-2-carboxylic acid
Thermo Scientific Chemicals

Azetidine-2-carboxylic acid

CAS: 2517-04-6 | C4H7NO2 | 101.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466900050
Price (EUR)
273,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
273,00
Each
Chemical Identifiers
CAS2517-04-6
IUPAC Nameazetidine-2-carboxylic acid
Molecular FormulaC4H7NO2
InChI KeyIADUEWIQBXOCDZ-UHFFFAOYNA-N
SMILESOC(=O)C1CCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Water=<1 %
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only