Myclobutanil
Thermo Scientific Chemicals

Myclobutanil

CAS: 88671-89-0 | C15H17ClN4 | 288.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467140010
Price (EUR)
215,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
215,00
Each
Chemical Identifiers
CAS88671-89-0
IUPAC Name2-(4-chlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]hexanenitrile
Molecular FormulaC15H17ClN4
InChI KeyHZJKXKUJVSEEFU-UHFFFAOYNA-N
SMILESCCCCC(CN1C=NC=N1)(C#N)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point69°C to 76°C
Water=<1 %
HPLC>=94 %
Appearance (Color)White to light yellow to light beige
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only