Ezetimibe, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ezetimibe, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 467692500
Price (EUR)
79,90
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
79,90
Each
Specifications
CAS163222-33-1
Chemical Name or MaterialEzetimibe
Formula Weight409.43
Molecular FormulaC24H21F2NO3
Quantity250 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only