2-Methylindene, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methylindene, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 467760010
Price (EUR)
63,20
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
63,20
Each
Chemical Identifiers
CAS2177-47-1
IUPAC Name2-methyl-1H-indene
Molecular FormulaC10H10
InChI KeyYSAXEHWHSLANOM-UHFFFAOYSA-N
SMILESCC1=CC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
Refractive index1.5665 to 1.5675 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only