3-Chloro-4-fluorophenylhydrazine hydrochloride, 98%
3-Chloro-4-fluorophenylhydrazine hydrochloride, 98%
3-Chloro-4-fluorophenylhydrazine hydrochloride, 98%
Thermo Scientific Chemicals

3-Chloro-4-fluorophenylhydrazine hydrochloride, 98%

CAS: 175135-74-7 | C6H7Cl2FN2 | 197.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A10175.06
also known as A10175-06
Price (EUR)
63,40
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
63,40
Each
Chemical Identifiers
CAS175135-74-7
IUPAC Name(3-chloro-4-fluorophenyl)hydrazine
Molecular FormulaC6H6ClFN2
InChI KeyDUORBNTWCCJANU-UHFFFAOYSA-N
SMILESNNC1=CC=C(F)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale brown or pale pink
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only