Thermo Scientific Chemicals

Diethyl benzylphosphonate, 99%, Thermo Scientific Chemicals

Catalog number: A10645.30
250 g, Each
Thermo Scientific Chemicals

Diethyl benzylphosphonate, 99%, Thermo Scientific Chemicals

Catalog number: A10645.30
250 g, Each
Quantity
Catalog number: A10645.30
also known as A10645-30
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
1080-32-6
IUPAC Name
diethyl benzylphosphonate
Molecular Formula
C11H17O3P
InChI Key
AIPRAPZUGUTQKX-UHFFFAOYSA-N
SMILES
CCOP(=O)(CC1=CC=CC=C1)OCC
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4950-1.4980 @ 20?C
Form
Liquid

Description

Diethyl benzylphosphonate is used as a reactant for synthesis of 3,5-dihydroxy-4-isopropylstilbene for treatment of skin disorders and natural cytotoxic marine products of polyketide origin via intramolecular Diels-Alder reactions. It is also used as reactant for cyclization of aryl ethers, amines and amides & for investigating the effects of functional groups on the performance of clue organic LEDs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl benzylphosphonate is used as a reactant for synthesis of 3,5-dihydroxy-4-isopropylstilbene for treatment of skin disorders and natural cytotoxic marine products of polyketide origin via intramolecular Diels-Alder reactions. It is also used as reactant for cyclization of aryl ethers, amines and amides & for investigating the effects of functional groups on the performance of clue organic LEDs.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing