L-Alanine ethyl ester hydrochloride, 98+%
L-Alanine ethyl ester hydrochloride, 98+%
L-Alanine ethyl ester hydrochloride, 98+%
Thermo Scientific Chemicals

L-Alanine ethyl ester hydrochloride, 98+%

CAS: 1115-59-9 | C5H12ClNO2 | 153.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11271.14
also known as A11271-14
Price (EUR)
156,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
156,00
Each
Chemical Identifiers
CAS1115-59-9
IUPAC Namehydrogen ethyl 2-aminopropanoate chloride
Molecular FormulaC5H12ClNO2
InChI KeyJCXLZWMDXJFOOI-UHFFFAOYNA-N
SMILES[H+].[Cl-].CCOC(=O)C(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Optical Rotation+2.9° ± 0.5° (c=2.5 in water)
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Melting Point (clear melt)74.0-81.0°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only