(3-Aminopropyl)trimethoxysilane, 97%
(3-Aminopropyl)trimethoxysilane, 97%
(3-Aminopropyl)trimethoxysilane, 97%
Thermo Scientific Chemicals

(3-Aminopropyl)trimethoxysilane, 97%

CAS: 13822-56-5 | C6H17NO3Si | 179.291 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A11284.22
also known as A11284-22
Price (EUR)
48,65
Online Exclusive
53,90
Save 5,25 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
48,65
Online Exclusive
53,90
Save 5,25 (10%)
Each
Chemical Identifiers
CAS13822-56-5
IUPAC Name(3-aminopropyl)trimethoxysilane
Molecular FormulaC6H17NO3Si
InChI KeySJECZPVISLOESU-UHFFFAOYSA-N
SMILESCO[Si](CCCN)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4205-1.4255 @ 20?C
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible withstrong acids, strong oxidizing agents, water, bases, alcohols and peroxides.
RUO – Research Use Only

General References:

  1. Schalken, J. R. G.; Creatore, M.; Verhoeven, P.; Stevens, A. mu Plasma Printing Deposition of Amine-Containing Organo-Silane Polymers by Means of 3-Aminopropyl Trimethoxysilane. Nanosci. Nanotechnol. Lett. 2015, 7 (1), 62-66.
  2. Jones, S.; Kolpin, A.; Tsang, S. C. E. Modification of Pd for formic acid decomposition by support grafted functional groups. Catal. Struct. React. 2015, 1 (1), 19-24.