2,5-Dihydroxy-1,4-benzoquinone, 98%
2,5-Dihydroxy-1,4-benzoquinone, 98%
2,5-Dihydroxy-1,4-benzoquinone, 98%
Thermo Scientific Chemicals

2,5-Dihydroxy-1,4-benzoquinone, 98%

CAS: 615-94-1 | C6H4O4 | 140.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11442.06
also known as A11442-06
Price (EUR)
31,50
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
31,50
Each
Chemical Identifiers
CAS615-94-1
IUPAC Name2,5-dihydroxycyclohexa-2,5-diene-1,4-dione
Molecular FormulaC6H4O4
InChI KeyQFSYADJLNBHAKO-UHFFFAOYSA-N
SMILESOC1=CC(=O)C(O)=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormPowder
Appearance (Color)Cream to yellow-orange to brown
Assay (HPLC)≥97.5%
2,5-dihydroxy-1,4-benzoquinone dianion, and squarate as bridging ligands in the synthesis and magnetic properties of binuclear iron(III) complexes with oxalate. A Dinuclear 2,5-Dihydroxy-1,4-benzoquinone acts as a bridging ligand Ruthenium(II) Complex with the Dianion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-dihydroxy-1,4-benzoquinone dianion, and squarate as bridging ligands in the synthesis and magnetic properties of binuclear iron(III) complexes with oxalate. A Dinuclear 2,5-Dihydroxy-1,4-benzoquinone acts as a bridging ligand Ruthenium(II) Complex with the Dianion.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Michael D. Ward. A Dinuclear Ruthenium(II) Complex with the Dianion of 2,5-Dihydroxy-1,4-benzoquinone as Bridging Ligand. Redox, Spectroscopic, and Mixed-Valence Properties. norg. Chem. 1996, 35 (6) ,1712-1714.
  2. Francesc Lloret,; Miguel Julve,; Juan Faus,; Xavier Solans,; Yves Journaux,; Irene Morgenstern-Badarau. Synthesis and magnetic properties of binuclear iron(III) complexes with oxalate, 2,5-dihydroxy-1,4-benzoquinone dianion, and squarate as bridging ligands. Crystal structure of (.mu.-1,3-squarato)bis[(N,N'-ethylenebis(salicylideneaminato))(methanol)iron(III)]. Journal name. 1990, 29 (12), 2232-2237.