p-Tolyl isothiocyanate, 97%
p-Tolyl isothiocyanate, 97%
p-Tolyl isothiocyanate, 97%
Thermo Scientific Chemicals

p-Tolyl isothiocyanate, 97%

CAS: 622-59-3 | C8H7NS | 149.211 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11499.14
also known as A11499-14
Price (EUR)
130,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
130,00
Each
Chemical Identifiers
CAS622-59-3
IUPAC Name1-isothiocyanato-4-methylbenzene
Molecular FormulaC8H7NS
InChI KeyABQKHKWXTUVKGF-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.6330-1.6380 @ 20?C
Appearance (Color)Clear colorless to yellow
p-Tolyl isothiocyanate has been used in the preparation of 6-[1-amino-3-(p-tolyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione. It is used as capping agent to cap the N-terminal ends of polypeptide. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolyl isothiocyanate has been used in the preparation of 6-[1-amino-3-(p-tolyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione. It is used as capping agent to cap the N-terminal ends of polypeptide. It is also used as pharmaceutical intermediate.

Solubility
Decompose in water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture.
RUO – Research Use Only

General References:

  1. Thorfinnur Gunnlaugsson; Paul E Kruger; Paul Jensen; Juliann Tierney; Haslin Dato Paduka Ali; Gillian M Hussey. Colorimetric naked eye sensing of anions in aqueous solution. Journal of Organic Chemistry.2005, 70 (26), 10875-10878.
  2. D H Swenson; K el-Bayoumy; G H Shuie; S S Hecht; E Fiala; F F Kadlubar; J P Freeman. Synthesis of N-(purin-8-yl)arylamines. Chemical Research in Toxicology.1993, 6 (4), 480-485.