3-Amino-4-hydroxybenzhydrazide, 98%, Thermo Scientific Chemicals
3-Amino-4-hydroxybenzhydrazide, 98%, Thermo Scientific Chemicals
3-Amino-4-hydroxybenzhydrazide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-4-hydroxybenzhydrazide, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
A11538.03
also known as A11538-03
1 g
Catalog number A11538.03
also known as A11538-03
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7450-57-9
IUPAC Name3-amino-4-hydroxybenzohydrazide
Molecular FormulaC7H9N3O2
InChI KeyKNKNIBYXWQUDIH-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=C(O)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Assay (HPLC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only