Triphenylphosphine dibromide, 96%
Triphenylphosphine dibromide, 96%
Triphenylphosphine dibromide, 96%
Triphenylphosphine dibromide, 96%
Thermo Scientific Chemicals

Triphenylphosphine dibromide, 96%

CAS: 1034-39-5 | C18H15Br2P | 422.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A11552.09
also known as A11552-09
Price (EUR)
35,70
キャンペーン価格
42,00
Save 6,30 (15%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (EUR)
35,70
キャンペーン価格
42,00
Save 6,30 (15%)
Each
Add to cart
Chemical Identifiers
CAS1034-39-5
IUPAC Namedibromotriphenyl-λ⁵-phosphane
Molecular FormulaC18H15Br2P
InChI KeyOCXGTPDKNBIOTF-UHFFFAOYSA-N
SMILESBrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder or fused solid
Assay (Titration ex Bromide)≥95.0 to ≤105.0% (U.K. specification material)
Assay from Supplier's CofA≥95.0% to ≤105.0% (U.S. specification material)
CommentPurchased in U.K. and U.S.
Triphenylphosphine dibromide is used as a reagent for conversion of aldoximes into nitriles. It acts as a reagent for cleavage of acetals. It reacts with amines to give iminophosphoranes. Further, it serves as a reagent in the preparation of alkyl and aryl bromides from alcohols and phenols without rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylphosphine dibromide is used as a reagent for conversion of aldoximes into nitriles. It acts as a reagent for cleavage of acetals. It reacts with amines to give iminophosphoranes. Further, it serves as a reagent in the preparation of alkyl and aryl bromides from alcohols and phenols without rearrangement.

Solubility
Soluble in water, dichloromethane, acetonitrile, phenyl cyanide, dimethyl formamide, ether, chloroform, alcohol, carbon tetrachloride, acetic acid and carbon disulfide. Sparingly soluble in phenyl chloride, triphenylphosphine and phenyl hydride. Insoluble in aqueous bromine.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong bases, strong oxidizing agents, bases and active metals.
RUO – Research Use Only

General References:

  1. Reagent for the conversion of alcohols and phenols to bromides in good yields: J. Org. Chem., 37, 626 (1972). Particularly useful for alcohols where sensitive functionality is present: J. Org. Chem., 49, 431 (1984); Synth. Commun., 22, 2945 (1992). Cleaves dialkyl ethers to the two alkyl bromides: J. Am. Chem. Soc., 86, 5037 (1964); J. Org. Chem., 37, 626 (1972).
  2. Reaction with amines gives iminophosphoranes, which have been used as a means of blocking an amino group to permit monoalkylation: Synthesis, 295 (1980). Iminophosphoranes also undergo a Wittig-like condensation with CO2 or CS2 to give the isocyanates or isothiocyanates respectively: Synthesis, 596 (1982):
  3. Ureas are dehydrated to carbodiimides: Liebigs Ann. Chem., 718, 24 (1968); Synthesis, 373 (1981).
  4. The reagent formed with DMSO at low temperatures provides an alternative to the Swern oxidation for selective conversion of alcohols to carbonyl compounds: Tetrahedron Lett., 43, 8355 (2002).
  5. Wang, R.; Zhao, Y.; Zhu, C.; Huang, X. New Synthesis of 3, 6-Dibromophthalonitrile and Phthalocyanine Having Eight Thienyl Substituents at Peripheral alpha-Positions. J. Heterocycl. Chem. 2015, 52 (4), 1230-1233.
  6. Koshizawa, T.; Yamazaki, Y.; Ohgiya, T.; Shibuya, K. A facile method for converting alcohol to thioether and its application in the synthesis of a novel GPR119 agonist. Tetrahedron 2015, 71 (21), 3231-3236.