Bromocyclohexane, 98%
Bromocyclohexane, 98%
Bromocyclohexane, 98%
Thermo Scientific Chemicals

Bromocyclohexane, 98%

CAS: 108-85-0 | C6H11Br | 163.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A11593.22
also known as A11593-22
Price (EUR)
28,30
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
28,30
Each
Chemical Identifiers
CAS108-85-0
IUPAC Namebromocyclohexane
Molecular FormulaC6H11Br
InChI KeyAQNQQHJNRPDOQV-UHFFFAOYSA-N
SMILESBrC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4940-1.4980 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to pale yellow
Bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air & Light Sensitive. Store away from strong bases.
RUO – Research Use Only

General References:

  1. Songhee Han; Hyun Sik Yoo; Sang Kyu Kim. Conformer-specific ionization spectroscopy of bromocyclohexane: equatorial versus axial conformers. Journal of Physical Chemistry A: Molecules, Spectroscopy, Kinetics, Environment and General Theory.2010, 114 (37), 10005-10010.
  2. Gagan Deep; Rahul Kumar; Anil K Jain; Chapla Agarwal; Rajesh Agarwal. Isolation of axial conformers of chloro- and bromocyclohexane in a pure state as inclusion complexes with 9,9'-bianthryl, and the discovery of a novel 1,3 diaxial Cl...H weak interaction..Chemical Communications.2005, (29), 3646-3648.