2-Mercaptopyridine, 98%
2-Mercaptopyridine, 98%
2-Mercaptopyridine, 98%
2-Mercaptopyridine, 98%
Thermo Scientific Chemicals

2-Mercaptopyridine, 98%

CAS: 2637-34-5 | C5H5NS | 111.162 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11741.14
also known as A11741-14
Price (EUR)
70,65
線上優惠
78,80
Save 8,15 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
70,65
線上優惠
78,80
Save 8,15 (10%)
Each
Add to cart
Chemical Identifiers
CAS2637-34-5
IUPAC Name1,2-dihydropyridine-2-thione
Molecular FormulaC5H5NS
InChI KeyWHMDPDGBKYUEMW-UHFFFAOYSA-N
SMILESS=C1NC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay (HPLC)≥97.5%
Assay (GC)≥97.5%
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
View more
Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.

Solubility
Slightly soluble in chloroform, methanol and dimethyl sulfoxide.

Notes
Air Sensitive, keep the container tightly sealed. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. David B Cordes; and Lyall R Hanton. Structural invariance in silver(I) coordination networks formed using flexible four-armed thiopyridine ligands. Inorganic Chemistry. 2007, 46 (5), 1634-1644.
  2. Alexey V Chuchuryukin et. al. Hydroxy- and mercaptopyridine pincer platinum and palladium complexes generated by silver-free halide abstraction. Inorganic Chemistry, 2006, 45 (5), 2045-2054.
  3. S-Acyl derivatives react with Grignard reagents in a high-yielding synthesis of ketones: J. Am. Chem. Soc., 95, 4763 (1973):
  4. Titanium enolates of S-acyl derivatives react with imines in a one pot synthesis of ß-lactams: J. Org. Chem., 57, 4155 (1992).