Cyclohexanone 2,4-dinitrophenylhydrazone, 99%
Cyclohexanone 2,4-dinitrophenylhydrazone, 99%
Cyclohexanone 2,4-dinitrophenylhydrazone, 99%
Thermo Scientific Chemicals

Cyclohexanone 2,4-dinitrophenylhydrazone, 99%

CAS: 1589-62-4 | C12H14N4O4 | 278.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A12203.14
also known as A12203-14
Price (EUR)
608,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
608,00
Each
Chemical Identifiers
CAS1589-62-4
IUPAC Name1-cyclohexylidene-2-(2,4-dinitrophenyl)hydrazine
Molecular FormulaC12H14N4O4
InChI KeyQLWXZRVOHCYKKK-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(NN=C2CCCCC2)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
FormPowder
Elemental AnalysisC: 51.02-52.58% (Theory: 51.8%)
Melting Point (clear melt)156.0-163.0?C
Elemental AnalysisH: 4.99-5.14% (Theory: 5.07%)
View more
Cyclohexanone 2,4-dinitrophenylhydrazone is a useful standard for elemental analysis. Also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanone 2,4-dinitrophenylhydrazone is a useful standard for elemental analysis. Also used as a pharmaceutical intermediate.

Solubility
Soluble in water (partly).

Notes
Store at 2-8°C. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Norbert Binding; Karsten Schilder; Peter A. Czeschinski; Ute Witting. Simultaneous determination of airborne acetaldehyde, acetone, 2-butanone, and cyclohexanone using sampling tubes with 2,4-dinitrophenylhydrazine-coated solid sorbent. Toxicology Letters. 1998, 96- 97, 289-299.
  2. Ariane Kahnt; Yoshiteru Iinuma; Olaf Böge; Anke Mutzel; Hartmut Herrmann. Denuder sampling techniques for the determination of gas-phase carbonyl compounds: A comparison and characterisation of in situ and ex situ derivatisation methods. Journal of Chromatography B. 2011, 879 (17-18), 1402-1411.