Ethyl coumarin-3-carboxylate, 98%
Ethyl coumarin-3-carboxylate, 98%
Ethyl coumarin-3-carboxylate, 98%
Thermo Scientific Chemicals

Ethyl coumarin-3-carboxylate, 98%

CAS: 1846-76-0 | C12H10O4 | 218.208 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A12553.03
also known as A12553-03
Price (EUR)
25,90
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
25,90
Each
Chemical Identifiers
CAS1846-76-0
IUPAC Nameethyl 2-oxo-2H-chromene-3-carboxylate
Molecular FormulaC12H10O4
InChI KeyXKHPEMKBJGUYCM-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC2=CC=CC=C2OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
Melting Point (clear melt)89.0-95.0?C
FormPowder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only