Methyl 4-methoxybenzoate, 98+%
Methyl 4-methoxybenzoate, 98+%
Methyl 4-methoxybenzoate, 98+%
Thermo Scientific Chemicals

Methyl 4-methoxybenzoate, 98+%

CAS: 121-98-2 | C9H10O3 | 166.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12598.22
also known as A12598-22
Price (EUR)
95,30
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
95,30
Each
Chemical Identifiers
CAS121-98-2
IUPAC Namemethyl 4-methoxybenzoate
Molecular FormulaC9H10O3
InChI KeyDDIZAANNODHTRB-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)45.0-52.0?C
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only