Thermo Scientific Chemicals

Nickel(II) acetate tetrahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A13026.36
500 g, Each
Thermo Scientific Chemicals

Nickel(II) acetate tetrahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A13026.36
500 g, Each
Quantity
Catalog number: A13026.36
also known as A13026-36
Price (EUR)
Price: 65,50
Online price: 58,95
Your price:
Quantity
-

Chemical Identifiers

CAS
6018-89-9
IUPAC Name
nickel(2+) diacetate tetrahydrate
Molecular Formula
C4H14NiO8
InChI Key
OINIXPNQKAZCRL-UHFFFAOYSA-L
SMILES
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O
Form
Crystals or powder or crystalline powder
Assay (unspecified)
≥98.0 to ≤102.0% (based on elemental Ni)
Complexometric Titration
Nickel Ni: 23.1-24.05%
Appearance (Color)
Green

Description

Nickel(II) acetate tetrahydrate is used in electroplating. It is also used in the preparation of polynuclear-nickel polyoxotungstate cluster compounds, which are used in designing molecular magnets. Further, it is used as a catalyst and mordant for dyeing textiles, as a sealant for anodized aluminum and to make other nickel compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nickel(II) acetate tetrahydrate is used in electroplating. It is also used in the preparation of polynuclear-nickel polyoxotungstate cluster compounds, which are used in designing molecular magnets. Further, it is used as a catalyst and mordant for dyeing textiles, as a sealant for anodized aluminum and to make other nickel compounds.

Solubility
Soluble in water and methanol. Insoluble in diethyl ether and n-octanol.

Notes
Hygroscopic. Incompatible with oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Respiratory Sensitization
    Category 1
    Skin Sensitization
    Category 1
    Germ Cell Mutagenicity
    Category 2
    Carcinogenicity
    Category 1A
    Reproductive Toxicity
    Category 1B
    Specific target organ toxicity - (repeated exposure)
    Category 1
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 + H332 - Harmful if swallowed or if inhaled

    H317 - May cause an allergic skin reaction

    H334 - May cause allergy or asthma symptoms or breathing difficulties if inhaled

    H341 - Suspected of causing genetic defects

    H350i - May cause cancer by inhalation

    H360D - May damage the unborn child

    H372 - Causes damage to organs through prolonged or repeated exposure

    H410 - Very toxic to aquatic life with long lasting effects

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P273 - Avoid release to the environment

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    Additional EU labelling

    Restricted to professional users